Learn More
Carbenoxolone disodium salt, 97+%
CAS: 7421-40-1 | C34H48Na2O7 | 614.73 g/mol
$109.97 - $414.85
Chemical Identifiers
| CAS | 7421-40-1 |
|---|---|
| Molecular Formula | C34H48Na2O7 |
| Molecular Weight (g/mol) | 614.73 |
| MDL Number | MFCD00079043 |
| InChI Key | BQENDLAVTKRQMS-SBBGFIFASA-L |
| Synonym | carbenoxolone disodium salt, disodium 3beta-3-3-carboxylatopropanoyl oxy-11-oxoolean-12-en-30-oate, disodium 4as,6as,6br,10s,12as,12br,14br-10-3-carboxylatopropanoyl oxy-2,4a,6a,6b,9,9,12a-heptamethyl-13-oxo-3,4,5,6,7,8,8a,10,11,12,12b,14b-dodecahydro-1h-picene-2-carboxylate |
| PubChem CID | 133687779 |
| IUPAC Name | disodium;(2R,4aR,6aS,6aR,6bS,8aS,10R,12aR,14bS)-10-(3-carboxylatopropanoyloxy)-2,4a,6a,6b,9,9,12a-heptamethyl-13-oxo-3,4,5,6,6a,7,8,8a,10,11,12,14b-dodecahydro-1H-picene-2-carboxylate |
| SMILES | [Na+].[Na+].[H][C@@]12C[C@](C)(CC[C@]1(C)CC[C@]1(C)C2=CC(=O)[C@]2([H])[C@@]3(C)CC[C@H](OC(=O)CCC([O-])=O)C(C)(C)[C@]3([H])CC[C@@]12C)C([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ6371403
|
Thermo Scientific Chemicals
J6371403 |
1 g |
Each for $109.97
|
|
|||||
|
AAJ6371406
|
Thermo Scientific Chemicals
J6371406 |
5 g |
Each for $414.85
|
|
|||||
Description
Carbenoxolone is anti-inflammatory. It is often used as a gap junction inhibitor in neuroscience research. Carbenoxolone is also an inhibitor of the steroid dehydrogenase enzymes 11 β-hydroxysteroid dehydrogenase (human) and 3α, 20 β-hydroxysteroid dehydrogenase (bacterial). It is used for the treatment of digestive tract ulcers, especially in the stomach. Used in the treatment of peptic, mainly gastric, ulcers. It is often used as a gap junction inhibitor in neuroscience research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
SolubilitySoluble in water to 100 mM.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from heat.
Chemical Identifiers
| 7421-40-1 | |
| 614.73 | |
| BQENDLAVTKRQMS-SBBGFIFASA-L | |
| 133687779 | |
| [Na+].[Na+].[H][C@@]12C[C@](C)(CC[C@]1(C)CC[C@]1(C)C2=CC(=O)[C@]2([H])[C@@]3(C)CC[C@H](OC(=O)CCC([O-])=O)C(C)(C)[C@]3([H])CC[C@@]12C)C([O-])=O |
| C34H48Na2O7 | |
| MFCD00079043 | |
| carbenoxolone disodium salt, disodium 3beta-3-3-carboxylatopropanoyl oxy-11-oxoolean-12-en-30-oate, disodium 4as,6as,6br,10s,12as,12br,14br-10-3-carboxylatopropanoyl oxy-2,4a,6a,6b,9,9,12a-heptamethyl-13-oxo-3,4,5,6,7,8,8a,10,11,12,12b,14b-dodecahydro-1h-picene-2-carboxylate | |
| disodium;(2R,4aR,6aS,6aR,6bS,8aS,10R,12aR,14bS)-10-(3-carboxylatopropanoyloxy)-2,4a,6a,6b,9,9,12a-heptamethyl-13-oxo-3,4,5,6,6a,7,8,8a,10,11,12,14b-dodecahydro-1H-picene-2-carboxylate |
Specifications
| 7421-40-1 | |
| C34H48Na2O7 | |
| 1 g | |
| carbenoxolone disodium salt, disodium 3beta-3-3-carboxylatopropanoyl oxy-11-oxoolean-12-en-30-oate, disodium 4as,6as,6br,10s,12as,12br,14br-10-3-carboxylatopropanoyl oxy-2,4a,6a,6b,9,9,12a-heptamethyl-13-oxo-3,4,5,6,7,8,8a,10,11,12,12b,14b-dodecahydro-1h-picene-2-carboxylate | |
| BQENDLAVTKRQMS-SBBGFIFASA-L | |
| disodium;(2R,4aR,6aS,6aR,6bS,8aS,10R,12aR,14bS)-10-(3-carboxylatopropanoyloxy)-2,4a,6a,6b,9,9,12a-heptamethyl-13-oxo-3,4,5,6,6a,7,8,8a,10,11,12,14b-dodecahydro-1H-picene-2-carboxylate | |
| 133687779 | |
| ≥97% | |
| Carbenoxolone disodium salt |
| White to Cream | |
| MFCD00079043 | |
| 14,1793 | |
| Soluble in water to 100 mM. | |
| [Na+].[Na+].[H][C@@]12C[C@](C)(CC[C@]1(C)CC[C@]1(C)C2=CC(=O)[C@]2([H])[C@@]3(C)CC[C@H](OC(=O)CCC([O-])=O)C(C)(C)[C@]3([H])CC[C@@]12C)C([O-])=O | |
| 614.73 | |
| 614.7 | |
| Solid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 231-044-0
RTECSNumber : RK0250000
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only