missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(Carbethoxymethylene)triphenylphosphorane, 98+%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 108295000

 View more versions of this product

Catalog No. AC108295000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

For C-C Bond Formation (Olefination)
Specifications

Specifications

(Carbethoxymethylene)triphenylphosphorane
98.0
Authentic
Glass bottle
(C6H5)3P=CHCO2C2H5
500g
carbethoxymethylene triphenylphosphorane, ethyl triphenylphosphoranylidene acetate, triphenylcarbethoxymethylenephosphorane, acetic acid, triphenylphosphoranylidene-, ethyl ester, ethyl 2-triphenylphosphoranylidene acetate, ethoxycarbonylmethylene triphenylphosphorane, carboethoxymethylidenetriphenylphosphorane, ethyl triphenylphosphoranilidene acetate, carbethoxytriphenylphosphonium methylide, carbethoxymethylidenetriphenylphosphorane
IIHPVYJPDKJYOU-UHFFFAOYSA-N
ethyl 2-(triphenyl-$l^{5}-phosphanylidene)acetate
70670
98+%
1099-45-2
100.0
98% min. (HPLC)
C22H21O2P
MFCD00009183
01,112; 02,60; 10,78
Solubility in water: insoluble
CCOC(=O)C=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
348.38
348.38

Chemical Identifiers

1099-45-2
348.38
IIHPVYJPDKJYOU-UHFFFAOYSA-N
70670
CCOC(=O)C=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
C22H21O2P
MFCD00009183
carbethoxymethylene triphenylphosphorane, ethyl triphenylphosphoranylidene acetate, triphenylcarbethoxymethylenephosphorane, acetic acid, triphenylphosphoranylidene-, ethyl ester, ethyl 2-triphenylphosphoranylidene acetate, ethoxycarbonylmethylene triphenylphosphorane, carboethoxymethylidenetriphenylphosphorane, ethyl triphenylphosphoranilidene acetate, carbethoxytriphenylphosphonium methylide, carbethoxymethylidenetriphenylphosphorane
ethyl 2-(triphenyl-$l^{5}-phosphanylidene)acetate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Toxic if swallowed.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 214-151-7

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only