Learn More
Carbon, activated, Norit ROW 0.8mm pellets, steam activated
CAS: 7440-44-0 | C | 12.011 g/mol
$70.78 - $395.42
Chemical Identifiers
| CAS | 7440-44-0 |
|---|---|
| Molecular Formula | C |
| Molecular Weight (g/mol) | 12.011 |
| MDL Number | MFCD00133992 |
| InChI Key | OKTJSMMVPCPJKN-UHFFFAOYSA-N |
| Synonym | graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve |
| PubChem CID | 5462310 |
| ChEBI | CHEBI:27594 |
| IUPAC Name | carbon |
| SMILES | [C] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL1633422
|
Thermo Scientific Chemicals
L1633422 |
100 g |
Each for $70.78
|
|
|||||
|
AAL1633436
|
Thermo Scientific Chemicals
L1633436 |
500 g |
Each for $115.63
|
|
|||||
|
AAL163340P
|
Thermo Scientific Chemicals
L163340P |
2000 g |
Each for $395.42
|
|
|||||
Description
Carbon, activated, Norit ROW 0.8mm pellets, steam activated is suitable for decolorization, deodorization and purification applications. It is mainly used for gas phase applications such as automotive emission control due to their low pressure drop, high mechanical strength and low dust content. It has high removal efficiency for removing degradation byproducts for drinking water. It also removes foaming causing impurities for the purification of amine and glycol process streams.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCarbon, activated, Norit ROW 0.8mm pellets, steam activated is suitable for decolorization, deodorization and purification applications. It is mainly used for gas phase applications such as automotive emission control due to their low pressure drop, high mechanical strength and low dust content. It has high removal efficiency for removing degradation byproducts for drinking water. It also removes foaming causing impurities for the purification of amine and glycol process streams.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with acids and oxidizing agents.
Chemical Identifiers
| 7440-44-0 | |
| 12.011 | |
| OKTJSMMVPCPJKN-UHFFFAOYSA-N | |
| 5462310 | |
| carbon |
| C | |
| MFCD00133992 | |
| graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve | |
| CHEBI:27594 | |
| [C] |
Specifications
| 7440-44-0 | |
| MFCD00133992 | |
| graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve | |
| OKTJSMMVPCPJKN-UHFFFAOYSA-N | |
| carbon | |
| 5462310 | |
| Carbon, activated, Norit ROW 0.8mm pellets, steam activated |
| C | |
| 100 g | |
| Insoluble in water. | |
| [C] | |
| 12.011 | |
| CHEBI:27594 |
Safety and Handling
H500
EINECSNumber : 231-153-3
RTECSNumber : FF5250100
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only