MilliporeSigma™ Supelco™ Carbon Adsorbent Sampler Kit, Matrix 20/40 GCB Kit
Supplier: MilliporeSigma™ Supelco™ 13027U
Specifications
Kit (5 Bottles) | |
20/40 Carbotrap(R) B (5 g); 20/40 Carbotrap C (5 g); 20/40 Carbotrap F (5 g); 20/40 Carbotrap x (5 g); 20/40 Carbotrap Y (5 g) |