missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Carbon Standard, (Sodium Carbonate-Sodium Bicarbonate), Certified, 1000ppm Inorganic (1mL = 1mg C), LabChem™
$49.91 - $72.09
Chemical Identifiers
CAS | 144-55-8 |
---|---|
Molecular Formula | CHNaO3 |
Molecular Weight (g/mol) | 84.01 |
MDL Number | MFCD00003528 |
InChI Key | UIIMBOGNXHQVGW-UHFFFAOYSA-M |
PubChem CID | 516892 |
ChEBI | CHEBI:32139 |
IUPAC Name | sodium hydrogen carbonate |
SMILES | [Na+].OC([O-])=O |
Chemical Identifiers
144-55-8 | |
84.01 | |
UIIMBOGNXHQVGW-UHFFFAOYSA-M | |
CHEBI:32139 | |
[Na+].OC([O-])=O |
CHNaO3 | |
MFCD00003528 | |
516892 | |
sodium hydrogen carbonate |
Specifications
1000ppm Inorganic (1mL = 1mg C) | |
Poly Bottle | |
99.21,0.44,0.35 | |
Passes Test | |
CHNaO3 | |
MFCD00003528 | |
UIIMBOGNXHQVGW-UHFFFAOYSA-M | |
[Na+].OC([O-])=O | |
84.01 | |
CHEBI:32139 | |
Certified |
144-55-8,497-19-8,144-55-8,7732-18-5 | |
125 mL | |
Colorless | |
1g/mL | |
Na2CO3 | |
Soluble in water | |
Liquid | |
sodium hydrogen carbonate | |
516892 | |
105.99 | |
Carbon Standard, 1000ppm, Inorganic (1mL = 1mg C), (Sodium Carbonate-Sodium Bicarbonate) |
Safety and Handling
GHS H Statement
Solution is not hazardous.
GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage : Room Temperature