missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Carbon tetrabromide, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 325155000

 View more versions of this product

Catalog No. AC325155000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
Specifications

Specifications

Carbon tetrabromide
88°C to 93°C
190°C
Plastic bottle
CBr4
500 g
15,352; 16,366
Solubility in water: insoluble
C(Br)(Br)(Br)Br
331.64
CHEBI:47875
98%
558-13-4
White
97.5% min. (On organic basis) (GC)
CBr4
MFCD00000117
01, 68
carbon tetrabromide, methane, tetrabromo, carbon bromide, methane tetrabromide, bromid uhlicity, carbontetrabromide, methane, tetrabromide, tetrabrommethan, bromid uhlicity czech
HJUGFYREWKUQJT-UHFFFAOYSA-N
tetrabromomethane
11205
331.64
Crystalline Solid

Chemical Identifiers

558-13-4
331.64
HJUGFYREWKUQJT-UHFFFAOYSA-N
11205
tetrabromomethane
CBr4
MFCD00000117
carbon tetrabromide, methane, tetrabromo, carbon bromide, methane tetrabromide, bromid uhlicity, carbontetrabromide, methane, tetrabromide, tetrabrommethan, bromid uhlicity czech
CHEBI:47875
C(Br)(Br)(Br)Br
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
May cause respiratory irritation.
Harmful if swallowed.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medi

GHS Signal Word: Warning

EINECSNumber : 209-189-6

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only