missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Carboplatin, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals J6043303

 View more versions of this product

Catalog No. AAJ6043303


Explore available promotions

Description

Description

Carboplatin is primarily used as an antitumor agent.Carboplatin is widely utilized as a chemotherapy drug to treat various types of cancer. It belongs to the group of platinum-based antineoplastic agents to interfere in DNA (deoxyribonucleic acid) repair. It is applied in combination other medications and used to inhibit cancer cell growth.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Specifications

Specifications

Carboplatin
White
[C4H6(CO2)2]Pt(NH3)2
1 g
carboplatin
BHKICZDKIIDMNR-UHFFFAOYSA-L
azanide;cyclobutane-1,1-dicarboxylate;platinum(4+)
122130863
Powder
41575-94-4
C6H12N2O4Pt
MFCD00070464
14,1822
Soluble in water.
N.N.O=C1O[Pt++]OC(=O)C11CCC1
371.26
371.25

Chemical Identifiers

41575-94-4
371.26
BHKICZDKIIDMNR-UHFFFAOYSA-L
122130863
N.N.O=C1O[Pt++]OC(=O)C11CCC1
C6H12N2O4Pt
MFCD00070464
carboplatin
azanide;cyclobutane-1,1-dicarboxylate;platinum(4+)
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H334-H340-H360-H302-H312-H332-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause genetic defects.
May damage fertility or the unborn child.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
May cause an allergic skin reaction.

P201-P202-P261-P264b-P270-P271-P272-P280g-P281-P285-P301+P312-P302+P352-P304+P340-P308+P313-P312-P330-P333+P313-P342+P311-P363-P501c

H302+H312+H332-H317-H334-H340-H350-H360

EINECSNumber : 255-446-0

RTECSNumber : TP2300000

TSCA : No

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only