missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Carbowax™ PEG 3350 (Powder), Fisher Chemical
Polyethylene Glycol
Supplier: Thermo Fisher Scientific P1463
Specifications
White | |
Solid | |
200°C | |
(C2H4O)n | |
1.2g/mL | |
3 kg | |
[H]OCCO | |
62.07 | |
Pass Test | |
00.000013 atm |
Carbowax™ PEG 3350 | |
25322-68-3 | |
54°C | |
MFCD01779601 | |
Polyethylene Glycol, Macrogol | |
LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
ethane-1,2-diol | |
3015 to 3685 | |
Poly Bottle |
Chemical Identifiers
25322-68-3 | |
62.07 | |
LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
ethane-1,2-diol |
(C2H4O)n | |
MFCD01779601 | |
Polyethylene Glycol, Macrogol | |
[H]OCCO |