missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Carbowax™ PEG 400 (NF), Fisher Chemical
Polyethylene Glycol
Supplier: Thermo Fisher Scientific P1671
Specifications
Carbowax™ PEG 401 | |
Liquid | |
8°C | |
(C2H4O)n | |
1.13g/mL | |
1 L | |
[H]OCCO | |
62.07 | |
NF | |
0.1% max. |
4.5 to 7.5 | |
25322-68-3 | |
380 to 420 | |
MFCD01779601 | |
Polyethylene Glycol, Macrogol, PEG | |
LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
ethane-1,2-diol | |
380 to 420 | |
Pass Test | |
Glass Bottle |
Chemical Identifiers
25322-68-3 | |
62.07 | |
LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
ethane-1,2-diol |
(C2H4O)n | |
MFCD01779601 | |
Polyethylene Glycol, Macrogol, PEG | |
[H]OCCO |