Carboxymethylcellulose Sodium, Low Viscosity, USP, Spectrum™ Chemical
CA193, 9004-32-4
Supplier:  Spectrum Chemical Mfg Cor CA193500GM
 View more versions of this product
Catalog No. 18601477
Explore available promotions
Description
Description
Spectrum™ Chemical Carboxymethylcellulose Sodium, Low Viscosity, Viscosity of 2 Percent Aqueous Solution @ 25 DEG C: 37.5 cP, USP is often used as a thickener and stabilizer. The USP grade indicates it is a grade suitable for personal care, cosmetic and pharmaceutical applications and were manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
9004-32-4
(C12 H14 O9 R6)n
500 g
[Na].CC(O)=O.OCC(O)C(O)C(O)C(O)C=O
263.20
10%
100%
MFCD00081472
DPXJVFZANSGRMM-UHFFFAOYNA-N
2,3,4,5,6-pentahydroxyhexanal acetic acid sodium
USP
Poly Bottle
Chemical Identifiers
9004-32-4
263.20
DPXJVFZANSGRMM-UHFFFAOYNA-N
[Na].CC(O)=O.OCC(O)C(O)C(O)C(O)C=O
(C12 H14 O9 R6)n
MFCD00081472
2,3,4,5,6-pentahydroxyhexanal acetic acid sodium