missing translation for 'onlineSavingsMsg'
Learn More

Carboxymethylcellulose Sodium, Medium Viscosity, USP, Spectrum™ Chemical

CA192, 9004-32-4

$403.00 - $26766.03

Chemical Identifiers

CAS 9004-32-4
Molecular Formula (C12 H14 O9 R6)n
Molecular Weight (g/mol) 263.20
MDL Number MFCD00081472
InChI Key DPXJVFZANSGRMM-UHFFFAOYNA-N
IUPAC Name 2,3,4,5,6-pentahydroxyhexanal acetic acid sodium
SMILES [Na].CC(O)=O.OCC(O)C(O)C(O)C(O)C=O
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18601481
View Documents
Spectrum Chemical Mfg Cor
CA192125GM
125 g Poly Bottle
Each for $403.00
Only null left
 
18601482
View Documents
Spectrum Chemical Mfg Cor
CA192500GM
500 g Poly Bottle
Each for $1,015.33
Only null left
 
18601483
View Documents
Spectrum Chemical Mfg Cor
CA19225KG
2.5 kg Poly Pail
Each for $2,697.73
Only null left
 
18601484
View Documents
Spectrum Chemical Mfg Cor
CA19212KG
12 kg Poly Pail
Each for $9,676.83
Only null left
 
18601485
View Documents
Spectrum Chemical Mfg Cor
CA19245KGBL
45 kg Fiber Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Spectrum™ Chemical Carboxymethylcellulose Sodium, Medium Viscosity, Viscosity of 2 Percent Aqueous Solution @ 25 DEG C: 400-800 cP, USP is often used as a thickener and stabilizer. The USP grade indicates it is a grade suitable for personal care, cosmetic and pharmaceutical applications and were manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications

Chemical Identifiers

9004-32-4
263.20
DPXJVFZANSGRMM-UHFFFAOYNA-N
[Na].CC(O)=O.OCC(O)C(O)C(O)C(O)C=O
(C12 H14 O9 R6)n
MFCD00081472
2,3,4,5,6-pentahydroxyhexanal acetic acid sodium

Specifications

9004-32-4
(C12 H14 O9 R6)n
125 g
[Na].CC(O)=O.OCC(O)C(O)C(O)C(O)C=O
263.20
10%
100%
MFCD00081472
DPXJVFZANSGRMM-UHFFFAOYNA-N
2,3,4,5,6-pentahydroxyhexanal acetic acid sodium
USP
Poly Bottle
Videos
SDS
Documents

Documents