Heathrow Scientific™ Cardboard Tube Storage Box Partition / Divider
This well partition is to be used with the Cardboard Tube Storage Box.
Supplier:  Heathrow Scientific™ HS2860T
 View more versions of this product
Catalog No. 17100448
Explore available promotions
Description
Description
  • Write-on surface for descriptions or self-numbering systems
  • Choose between two arrays: 4 x 4 or 6 x 6
Specifications
Specifications
White
Square
15 mL Tubes
6 x 6
Cardboard
Box Divider
36 Tubes
-196°C
Product Suggestions
Product Suggestions