Cargille Immersion Oil Edge
Edge
Greatly increases light-gathering power with high-power objectives
Supplier:  Cargille 16482A4
 View more versions of this product
Catalog No. 12-370A
Edge
Explore available promotions
Description
Description
Greatly increases light-gathering power with high-power objectives
  • Formulated to give refractive index of 1.5150 ± 0.0002 for D line at 23°C (73°F)
  • Nondrying, so index will not change when exposed to air
  • Contains no polychlorinated biphenyl (PCB) compounds
  • Conforms to ISO 8036/1 specifications
  • Standard for all oil-immersion microscopy
  • Type A oil has low viscosity and low fluorescence
  • Type B is a high-viscosity oil used in biomedical microscopy
  • Type NVH is a high-viscosity oil that meets requirements for horizontal, inverted and inclined instruments and projection equipment
  • Type FF oil is virtually nonfluorescent
  • Type DF is a very low-fluorescence oil for fluorescence microscopy with very high resolution
Specifications
Specifications
150 cSt
0.923g/cm3
Increases light-gathering power with high-power objectives
1.5150 ±0.0002
ISO 8036/1
Type A Immersion Oil
4 oz.