missing translation for 'onlineSavingsMsg'
Learn More

Carnauba wax, refined, No. 1, yellow

CAS: 8015-86-9

$81.35 - $198.00

Chemical Identifiers

CAS 8015-86-9
MDL Number MFCD00130724
Synonym Brazil wax
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC222382500
SDS View Documents
Thermo Scientific Chemicals
222382500
250 g Plastic bottle
Each for $81.35
Only null left
 
AC222380010
SDS View Documents
Thermo Scientific Chemicals
222380010
1 kg Plastic bottle
Each for $198.00
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

8015-86-9
MFCD00130724
Brazil wax

Specifications

2 to 7 (mg KOH/g)
81.0°C to 86.0°C
282°C
Plastic bottle
250 g
15,1846
Solubility in water: insoluble. Other solubilities: soluble in boiling alcohol,diethyl ether,benzene,,toluene,chloroform,ethyl acetate,turpentine,,petroleum ether,carbon disulfide;,soluble in alkalies
Carnauba wax
8015-86-9
Yellow
0.25% max.
MFCD00130724
78 to 95mg KOH/g
Brazil wax
Powder or Flakes
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 232-399-4

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only