missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ 6000 Series Cartridges
NIOSH-approved cartridges for use with 3M™ full and half facepiece respirators 6000, 7000 and FF-400 Series.
Supplier: 3M™ 7000051849
Description
NIOSH-approved cartridges for use with 3M™ full and half facepiece respirators 6000, 7000 and FF-400 Series.
- For use with 3M full and half facepiece respirators 6000, 7000 and FF-400 Series
- Bayonet cartridge connection
- NIOSH-approved for protection against a variety of gases, vapors, and particulate hazards*
- For contaminants up to 10X the Permissible Exposure Limit (PEL) with half-facepiece respirators or 50X the PEL (when fit tested) with full-facepiece respirators or according to specific OSHA standards or applicable government regulations, whichever is lower
- Not intended for use in atmospheres that are unknown or immediately dangerous to life or death (IDLH)
- Do not exceed maximum use concentrations established by regulatory standards
- These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product
- Misuse may result in sickness or death
*Details found at individual product level according
Specifications
| Organic Vapors, Acid Gases, Chlorine, Hydrogen Chloride, Sulfur Dioxide, Chlorine Dioxide, Hydrogen Sulfide, Hydrogen Fluoride | |
| Bayonet | |
| Cartridge | |
| 3M™ half and full facepiece 6000, 7000, ff-400 series respirators | |
| 60/Cs. | |
| 10.6 cm | |
| 8.6 cm |
| NIOSH-approved | |
| Yellow | |
| Assembly, cleaning, demolition, facility maintenance, machine operation, painting | |
| 6003 | |
| 4.2 in. | |
| 3.4 in. |