missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Specifications
Carvacrol | |
Yellow | |
C10H14O | |
100 g | |
RECUKUPTGUEGMW-UHFFFAOYSA-N | |
2-methyl-5-(propan-2-yl)phenol | |
10364 | |
150.22 | |
Liquid |
499-75-2 | |
238°C | |
MFCD00002236 | |
carvacrol, 5-isopropyl-2-methylphenol, isopropyl-o-cresol, o-thymol, antioxine, isothymol, karvakrol, 5-isopropyl-o-cresol, 2-p-cymenol, 2-hydroxy-p-cymene | |
CC(C)C1=CC=C(C)C(O)=C1 | |
150.22 | |
CHEBI:3440 | |
≥98.0% (GC) |
Chemical Identifiers
499-75-2 | |
150.22 | |
RECUKUPTGUEGMW-UHFFFAOYSA-N | |
10364 | |
2-methyl-5-(propan-2-yl)phenol |
C10H14O | |
MFCD00002236 | |
carvacrol, 5-isopropyl-2-methylphenol, isopropyl-o-cresol, o-thymol, antioxine, isothymol, karvakrol, 5-isopropyl-o-cresol, 2-p-cymenol, 2-hydroxy-p-cymene | |
CHEBI:3440 | |
CC(C)C1=CC=C(C)C(O)=C1 |
Safety and Handling
EINECSNumber : (4)-0057
RTECSNumber : FI1225000
TSCA : Yes