Learn More
(-)-Carveol, mixture of isomers, 97%, Thermo Scientific Chemicals
$91.52 - $1025.66
Chemical Identifiers
CAS | 99-48-9 |
---|---|
Molecular Formula | C10H16O |
Molecular Weight (g/mol) | 152.24 |
MDL Number | MFCD00869995,MFCD00062993 |
InChI Key | BAVONGHXFVOKBV-UHFFFAOYNA-N |
Synonym | carveol, p-mentha-6,8-dien-2-ol, p-mentha-1,8-dien-6-ol, 1-methyl-4-isopropenyl-6-cyclohexen-2-ol, 2-cyclohexen-1-ol, 2-methyl-5-1-methylethenyl, p-mentha-1 6 ,8-dien-2-ol, 5-isopropenyl-2-methyl-2-cyclohexen-1-ol, --carveol, l-carveol, 2-methyl-5-1-methylethenyl-2-cyclohexen-1-ol |
PubChem CID | 7438 |
ChEBI | CHEBI:23046 |
IUPAC Name | 2-methyl-5-prop-1-en-2-ylcyclohex-2-en-1-ol |
SMILES | CC(=C)C1CC=C(C)C(O)C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2063609
|
Thermo Scientific Chemicals
B2063609 |
10 g |
Each for $91.52
|
|
|||||
AAB2063618
|
Thermo Scientific Chemicals
B2063618 |
50 g |
Each for $250.53
|
|
|||||
AAB2063630
|
Thermo Scientific Chemicals
B2063630 |
250 g |
Each for $1,025.66
|
|
|||||
Description
It is a terpene that inhibits mammary carcinogenesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is a terpene that inhibits mammary carcinogenesis.
Solubility
Partly soluble in water. Soluble in alcohol, ethanol and DMSO.
Notes
Store at room temperature. Incompatible with strong oxidizing agents.
Chemical Identifiers
99-48-9 | |
152.24 | |
BAVONGHXFVOKBV-UHFFFAOYNA-N | |
7438 | |
2-methyl-5-prop-1-en-2-ylcyclohex-2-en-1-ol |
C10H16O | |
MFCD00869995,MFCD00062993 | |
carveol, p-mentha-6,8-dien-2-ol, p-mentha-1,8-dien-6-ol, 1-methyl-4-isopropenyl-6-cyclohexen-2-ol, 2-cyclohexen-1-ol, 2-methyl-5-1-methylethenyl, p-mentha-1 6 ,8-dien-2-ol, 5-isopropenyl-2-methyl-2-cyclohexen-1-ol, --carveol, l-carveol, 2-methyl-5-1-methylethenyl-2-cyclohexen-1-ol | |
CHEBI:23046 | |
CC(=C)C1CC=C(C)C(O)C1 |
Specifications
99-48-9 | |
227°C | |
C10H16O | |
MFCD00869995,MFCD00062993 | |
6269416 | |
Partly soluble in water. Soluble in alcohol,ethanol and DMSO. | |
CC(=C)C1CC=C(C)C(O)C1 | |
152.24 | |
CHEBI:23046 | |
97% |
0.96 | |
98°C (209°F) | |
1.496 | |
10 g | |
carveol, p-mentha-6,8-dien-2-ol, p-mentha-1,8-dien-6-ol, 1-methyl-4-isopropenyl-6-cyclohexen-2-ol, 2-cyclohexen-1-ol, 2-methyl-5-1-methylethenyl, p-mentha-1 6 ,8-dien-2-ol, 5-isopropenyl-2-methyl-2-cyclohexen-1-ol, --carveol, l-carveol, 2-methyl-5-1-methylethenyl-2-cyclohexen-1-ol | |
BAVONGHXFVOKBV-UHFFFAOYNA-N | |
2-methyl-5-prop-1-en-2-ylcyclohex-2-en-1-ol | |
7438 | |
152.24 | |
(-)-Carveol, mixture of isomers |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H320-H335
EINECSNumber : 202-757-4
RTECSNumber : OS8400000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only