missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Cary-Blair with Indicator Single Vial
Collect, preserve, and recover enteric bacterial pathogens with a fecal transport that comes with industry color-coding and sporked caps and is stored at room temperature.
$493.54 - $1594.31
Spécifications
| Product Type | Fecal Transport Vial |
|---|---|
| Fill Volume | 15 mL |
| Media Type | Cary-Blair |
| Packaging | Single vial. Packaged in partitioned boxes. |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Closure Color | Prix | Quantité | ||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Closure Color | Prix | Quantité | ||||
|
R21610
|
Thermo Scientific™
RE21610 |
12 Vials/Pk. | Green |
caisse de 12 chaque for $493.54
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||
|
R21925
|
Thermo Scientific™
R21925 |
120 Vials/Pk. | Green |
caisse de 120 chaque for $1,594.31
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||
Description
Collect, preserve, and recover enteric bacterial pathogens in stool specimens with Thermo Scientfic™ Cary-Blair with Indicator Single Vial, part of a complete collection of quality fecal transports which come with industry standard color-coding and easy-to-use sporked caps.
Maintain the viability of enteric bacterial pathogens during transport and storage with a formulation that includes 1.16% agar and is ready for use without any special preparation. Save valuable storage space with a transport that is stored in its original container at room temperature.- Cary-Blair with Indicator is an isotonic and nonnutritive medium, which does not contain a fermentable carbohydrate.
- Sodium thioglycollate is added to impede oxidation.
- Disodium phosphate is incorporated in the medium to act as a buffering agent, which prevents overgrowth of Escherichia coli and Enterobacter species.
- The relatively high pH minimizes the destruction of bacteria due to acid formation.
- Phenol red, a pH indicator, is added to demonstrate acidic shifts that can affect the viability of some enteric pathogens.
Add convenience and safety to your fecal collection procedures with a broad line of transport products which have a long shelf life and are designed especially for enteric diagnostic testing.
Look to us for convenient kit packaging options, including bulk kits.
Multilingual patient collection instructions are included.
Choose from a wide range of collection and preservation products for fecal specimens, which is important to the successful growth, isolation and detection of microorganisms.
- Cary Blair with Indicator
- Formalin 10%
- Formalin 5%
- PVA Modified Fixative
- SAF Fixative
- Zinc PVA Fixative
- Empty Vial
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Spécifications
| Fecal Transport Vial | |
| 15 mL | |
| Cary-Blair | |
| Single vial. Packaged in partitioned boxes. |