missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific Chemicals Caspase-3 Substrate III, fluorogenic
For cellular and molecular biology applications
$284.40 - $284.40
Specifications
Color | White to off-white |
---|---|
Form | Lyophilized |
Substrate Type | Caspase Substrate |
Molecular Weight (g/mol) | 1155.08 |
Storage Requirements | -30° to -10°C |
Catalog Number | Mfr. No. | Substrate Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Substrate Quantity | Price | Quantity | |||||
AAJ66881EXD
|
Thermo Scientific Chemicals
J66881EXD |
0.5mg |
Each for $284.40
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
White to off-white | |
Caspase Substrate | |
-30° to -10°C |
Lyophilized | |
1155.08 |
RUO – Research Use Only