Promega Caspase-Glo™ 3/7 Assay Kit DFS Item
Caspase-Glo™ 3/7 Assay is a homogeneous luminescent assay that measures caspase-3/7 activities
Supplier:  Promega G8092
 View more versions of this product
Catalog No. PR-G8092
Explore available promotions
Includes: Caspase-Glo 3/7 Substrate; Caspase-Glo 3/7 Buffer
Description
Description
  • Apoptosis Detection with Unmatched Speed and Sensitivity
  • High sensitivity—requires fewer cells and less enzyme
  • Simple add-mix-measure protocol—no sample preparation
  • Easily scalable to 96-, 384- and 1,536-well plate formats
  • Patent pending.
  • The method of recombinant expression of Coleoptera luciferase is covered by U.S. Patent Nos. 5,583,024; 5,674,713; and 5,700,673.
Specifications
Specifications
100 mL
0.25 to 1 hr.
96- or 384-well plates using either purified enzyme or cells in culture as sample
1 x 100mL Caspase-Glo™ 3/7 Buffer, 1 x 1 bottle Caspase-Glo™ 3/7 substrate
-30°C to -10°C
Liquid
Product Suggestions
Product Suggestions