Learn More
Invitrogen™ Cassette Clamp, left
Cassette clamps are replacement parts for the Mini Gel Tank. They lock and seal the gel cassettes within the Gel Runner Tank. The orientation is determined when facing the front of the gel tank.
Supplier: Invitrogen™ A25946
Description
About the Mini Gel TankThe Mini Gel Tank is compatible with a variety of Novex™ gels, NuPAGE™ gels, and Bolt™ Bis-Tris Plus gels. Each Mini Gel Tank can accommodate up to two gels per run. The unique tank design enables convenient side-by-side gel loading and enhanced viewing during use. Optimized conditions using constant voltage allow for Bolt™ Bis-Tris Plus gels to be run in approximately 35 minutes. Run times may vary depending on buffer conditions and power supplies being used.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
| Universal | |
| Mini | |
| 1 unit |
| Novex™ Mini Gels | |
| Cassette Clamp | |
| Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.