Fischer Technical Company™ Cast Iron High Vacuum Pump Plate
Cast Iron High Vacuum Pump Plate is high quality for years of classroom use.
$1123.18 - $1123.18
Catalog Number | Mfr. No. | Includes | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Includes | Price | Quantity | |||||
S41330-1
|
Fischer Technical Company™
GA100 |
Barbed Hose Connection, Vacuum Release Valve, Rubber Gasket |
Each for $1,123.18
|
|
|||||
Description
- Made of steel with rubber gasket
- Large enough to accommodate bell jars up to 9 in. (22.5 cm) in diameter
- Nickel-plated, 0.5 in. (1.25 cm) steel plate provides extra strength and is ground flat for a good seal
- Complete with a barbed hose connection, vacuum release valve and rubber gasket to provide a tight seal
- Replacement Gaskets Available
Specifications
Vacuum Plate | |
Classroom Use |