missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Supertek™ Cast Iron Support Ring
Cast iron support ring with side arm and V clamp to fit rods up to 13 mm. The distance from the support clamp center to the center of the ring is 4.5 in. for all rings.
$17.74 - $30.74
Catalog Number | Mfr. No. | Diameter (Metric) Inner | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (Metric) Inner | Price | Quantity | |||||
S012348
|
Supertek™
CH123711 |
40 mm |
Each for $22.18
|
|
|||||
S012347
|
Supertek™
CH123712 |
60 mm |
Each for $25.41
|
|
|||||
S012346
|
Supertek™
CH123713 |
78 mm |
Each for $30.74
|
|
|||||
S012345
|
Supertek™
CH123714 |
90 mm |
Each for $17.74
|
|
|||||
Description
- Cast iron support ring
- With side arm and V clamp to fit rods up to 13 mm
- The distance from the support clamp center to the center of the ring is 4.5 in.
Specifications
Cast Iron | |
18 mm | |
Circle | |
1 Ea. |