missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Catechol, 99+%, Thermo Scientific Chemicals

$59.58 - $451.98

Chemical Identifiers

CAS 120-80-9
Molecular Formula C6H6O2
Molecular Weight (g/mol) 110.11
MDL Number MFCD00002188
InChI Key YCIMNLLNPGFGHC-UHFFFAOYSA-N
Synonym pyrocatechol, catechol, 1,2-dihydroxybenzene, 1,2-benzenediol, pyrocatechin, 2-hydroxyphenol, o-benzenediol, pyrocatechine, o-dihydroxybenzene, o-dioxybenzene
PubChem CID 289
ChEBI CHEBI:18135
IUPAC Name benzene-1,2-diol
SMILES OC1=CC=CC=C1O
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC158980050
SDS View Documents
Thermo Scientific Chemicals
158980050
5 g Glass bottle N/A
 
AC158981000
SDS View Documents
Thermo Scientific Chemicals
158981000
100 g Glass bottle N/A
 
AC158985000
SDS View Documents
Thermo Scientific Chemicals
158985000
500 g Glass bottle N/A
 
AC158980025
SDS View Documents
Thermo Scientific Chemicals
158980025
2.5 kg Plastic Bottle N/A
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Specifications

Chemical Identifiers

120-80-9
110.11
YCIMNLLNPGFGHC-UHFFFAOYSA-N
289
benzene-1,2-diol
C6H6O2
MFCD00002188
pyrocatechol, catechol, 1,2-dihydroxybenzene, 1,2-benzenediol, pyrocatechin, 2-hydroxyphenol, o-benzenediol, pyrocatechine, o-dihydroxybenzene, o-dioxybenzene
CHEBI:18135
OC1=CC=CC=C1O

Specifications

120-80-9
Brown-Gray to White
131°C
99% min. (GC)
C6H6O2
5 g
pyrocatechol, catechol, 1,2-dihydroxybenzene, 1,2-benzenediol, pyrocatechin, 2-hydroxyphenol, o-benzenediol, pyrocatechine, o-dihydroxybenzene, o-dioxybenzene
YCIMNLLNPGFGHC-UHFFFAOYSA-N
benzene-1,2-diol
289
CHEBI:18135
99+%
Catechol
103.0°C to 106.0°C
245.0°C
Authentic
Glass bottle
MFCD00002188
15,8111
Solubility in water: 430g/L (20°C). Other solubilities: soluble in alcohol, benzene, chloroform, ether, and pyridine and aqueous solutions
OC1=CC=CC=C1O
110.11
2.38 mm2/s (110°C)
110.11
Crystalline Flakes
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Suspected of causing genetic defects.
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye damage.<b

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
If skin irritation occurs: Get medical advice/attention.
IF IN EYES: Rinse cau

GHS Signal Word: Danger

EINECSNumber : 204-427-5

Recommended Storage : Product may darken during storage

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only