missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific Chemicals Catechol Violet

Supplier:  Thermo Scientific Chemicals B2351314

 View more versions of this product

Catalog No. AAB2351314


Explore available promotions

Description

Description

Catechol Violet is used as a complexometric indicator dye. It inhibits the synthesis of globin in reticulocyte lysates. It is also used as a pH indicator with a transition interval from 0 to 8.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Catechol Violet is used as a complexometric indicator dye. It inhibits the synthesis of globin in reticulocyte lysates. It is also used as a pH indicator with a transition interval from 0 to 8.

Solubility
Soluble in water, ethylene glycol, monomethyl ether and ethanol.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

Catechol Violet
C19H14O7S
353938
RRRCKIRSVQAAAS-UHFFFAOYSA-N
4-[3-(3,4-dihydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]benzene-1,2-diol
66993
25 g
115-41-3
MFCD00005868
Soluble in water,ethylene glycol,monomethyl ether and ethanol.
OC1=CC=C(C=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C(O)=C1
386.37
386.38

Chemical Identifiers

115-41-3
386.37
RRRCKIRSVQAAAS-UHFFFAOYSA-N
4-[3-(3,4-dihydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]benzene-1,2-diol
C19H14O7S
MFCD00005868
66993
OC1=CC=C(C=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C(O)=C1
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335

P260-P261-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P363-P501c

H314-H335

EINECSNumber : 204-088-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only