missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific Chemicals Cathepsin B Inhibitor III
Supplier: Thermo Scientific Chemicals J65347LB0
Description
CA-074 is a potent and specific inhibitor of cathepsin B in vitro.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
5 mg | |
-30°C to -10°C | |
134448-10-5 | |
383.44g/mol | |
Soluble in methanol at 1mg/ml; Also soluble in DMSO or ethanol | |
Powder | |
CCCNC(=O)[C@H]1OC1C(=O)N[C@@H]([C@@H](C)CC)C(=O)N1CCC[C@H]1C(O)=O | |
383.45 |
White | |
An inhibitor of Cathepsin B | |
C18H29N3O6 | |
CA-074; [L-3-trans-(Propylcarbamoyl)oxirane-2-carbonyl]-L-isoleucyl-L-proline | |
Signal Transduction Reagent | |
ZEZGJKSEBRELAS-KSFNBINOSA-N | |
(2S)-1-[(2S,3S)-3-methyl-2-{[(3S)-3-(propylcarbamoyl)oxiran-2-yl]formamido}pentanoyl]pyrrolidine-2-carboxylic acid |
Chemical Identifiers
134448-10-5 | |
383.45 | |
CA-074; [L-3-trans-(Propylcarbamoyl)oxirane-2-carbonyl]-L-isoleucyl-L-proline | |
CCCNC(=O)[C@H]1OC1C(=O)N[C@@H]([C@@H](C)CC)C(=O)N1CCC[C@H]1C(O)=O |
C18H29N3O6 | |
ZEZGJKSEBRELAS-KSFNBINOSA-N | |
(2S)-1-[(2S,3S)-3-methyl-2-{[(3S)-3-(propylcarbamoyl)oxiran-2-yl]formamido}pentanoyl]pyrrolidine-2-carboxylic acid |
Safety and Handling
Recommended Storage : Store at -20°C
RUO – Research Use Only