missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD154 (CD40 Ligand) Monoclonal Antibody (24-31), FITC, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

Supplier:  Thermo Fisher Scientific 11154841

 View more versions of this product

Catalog No. 509776


Explore available promotions

Description

Description

Description: The 24-31 monoclonal antibody reacts with human CD154, a 39 kDa transmembrane glycoprotein also known as gp39 and CD40 ligand (CD40L). CD154 is a member of the TNF superfamily and is expressed transiently by activated T cells. Through its binding to CD40 on antigen presenting cells including B cells, monocytes/macrophages and dendritic cells, CD154 serves a crucial function in T-APC cognate interaction. CD154 interaction with CD40 transduces signals for T-dependent B-cell activation and induces B cell cycle entry. 24-31 cross-reacts with the cynomolgus monkey CD154. Applications Reported: 24-31 has been reported for use in flow cytometric analysis. Applications Tested: This 24-31 antibody has been pre-titrated and tested by flow cytometric analysis of stimulated normal human peripheral blood cells. This can be used at 5 μL (1 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.

CD40 Ligand (CD40-L), or CD154, is a membrane glycoprotein and differentiation antigen expressed on the surface of T cells. The CD40 Ligand stimulates B cell proliferation and secretion of all immunoglobulin isotypes in the presence of cytokines. CD40 Ligand has been shown to induce cytokine production and tumoricidal activity in peripheral blood monocytes. It also co-stimulates proliferation of activated T cells and this is accompanied by the production of IFN-gamma, TNF-alpha, and IL-2.
Specifications

Specifications

CD154 (CD40 Ligand)
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P29965
CD40LG
Affinity chromatography
RUO
102138630, 959
4° C, store in dark, DO NOT FREEZE!
Liquid
Flow Cytometry
24-31
FITC
CD40LG
RP23-153G22.3, CD154, CD40-L, Cd40l, HIGM1, IGM, IMD3, Ly-62, Ly62, T-BAM, TRAP, Tnfsf5, gp39, T-cell antigen Gp39, TNF-related activation protein, tumor necrosis factor ligand superfamily member 5
Mouse
25 Tests
Primary
Cynomolgus Monkey, Human
Antibody
IgG1 κ
Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only.