missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
CD54 (ICAM-1) Monoclonal Antibody (HA58), PerCP-eFluor™ 710, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$554.00 - $554.00
Specifications
Antigen | CD54 (ICAM-1) |
---|---|
Clone | HA58 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | PerCP-eFluor 710 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
501130758
|
Invitrogen
46054942 |
100 Tests |
Each for $554.00
|
|
||||||
Description
ICAM-1 (intercellular adhesion molecule) has 7 potential N-linked glycosylation sites. It is a single chain glycoprotein of Ig supergene family, present on unstimulated endo-thelial cells (EC) and on a variety of other cell types including activated fibroblasts, EC, macrophages, and lymphocytes. ICAM-1 mediates cell adhesion by binding to integrins CD11a/CD18 (leukocyte adhesion molecule, LFA-1) and to CD11b/CD18 (Mac-1).
ICAM-1 (CD54) is an 85-110 kDa single-chain type 1 integral membrane glycoprotein with an extracellular domain of five immunoglobulin superfamily repeats, a transmembrane region and a cytoplasmic domain. ICAM-1 has 7 potential N-linked glycosylation sites and shares considerable amino acid sequence homology with ICAM-3 (CD50) and ICAM-2 (CD102). ICAM-1 binds to integrins of type CD11a/CD18 (leukocyte adhesion molecule, LFA-1), or CD11b/CD18 (Mac-1) and is exploited by Rhinovirus as a receptor. ICAM-1 is expressed by activated endothelial cells and detected on epithelial cells, fibroblasts, chondrocytes, B lymphocytes, T lymphocytes (low), monocytes, macrophages, dendritic cells and neutrophils, with lower levels that increase upon inflammation. ICAM-1 is also detected in some carcinoma and melanoma cells. Soluble ICAM-1 is detectable in the plasma and is elevated in patients with various inflammatory syndromes.Specifications
CD54 (ICAM-1) | |
Flow Cytometry | |
PerCP-eFluor 710 | |
Mouse | |
Human | |
3383 | |
IgG1 κ | |
Affinity chromatography | |
ICAM1 |
HA58 | |
Monoclonal | |
Liquid | |
RUO | |
P05362 | |
ICAM1 | |
Primary | |
Antibody |
For Research Use Only.