Vileda Professional™ CE UltraSpeed Pro™ Double Bucket System
The most effective and easy to use flat mop bucket and wringer system for use on floors, walls and ceilings within Controlled Environments!
Supplier:  Vileda Professional™ 157775
Catalog No. 19-200-770
Explore available promotions
Description
Description
Demands on effectiveness, cost, ease of handling and staff training is constantly increasing. Whatever the cleaning requirements, the CE UltraSpeed Pro™ Double Bucket System will dramatically increase speed and cleaning performance while decreasing total costs and providing versatility and ergonomics.
  • Double bucket systems includes two buckets with flat mop wringer and chassis
  • Lightweight double bucket system allows for separation of dirty liquid and clean liquid minimizing cross-contamination
  • Touchless wringing of mops using the patented, fast and efficient wringer/press
  • Small and compact chassis (only 2.29 sq ft) means minimal storage space needed
  • Changeable color-coding clips within the buckets (red, blue, yellow, green)
  • Completely autoclavable with all stainless steel parts
  • Highly durable and withstands all common disinfectants
Specifications
Specifications
Autoclavable