Learn More
Cefazolin sodium salt, Thermo Scientific Chemicals
Inhibits bacterial cell wall synthesis
$219.96 - $867.64
Chemical Identifiers
CAS | 27164-46-1 |
---|---|
Molecular Formula | C14H16N8NaO4S3 |
Molecular Weight (g/mol) | 479.504 |
MDL Number | MFCD00056883 |
InChI Key | MTIAAUXSENDLGW-SLNAEPSVSA-N |
Synonym | Cefamedin |
PubChem CID | 131673922 |
IUPAC Name | (6R,7R)-3-[(5-methyl-1,3,4-thiadiazol-2-yl)sulfanylmethyl]-8-oxo-7-[[2-(tetrazol-1-yl)acetyl]amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;molecular hydrogen;sodium |
SMILES | [HH].CC1=NN=C(S1)SCC2=C(N3C(C(C3=O)NC(=O)CN4C=NN=N4)SC2)C(=O)O.[Na] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ6527403
|
Thermo Scientific Chemicals
J6527403 |
1 g |
Each for $219.96
|
|
|||||
AAJ6527406
|
Thermo Scientific Chemicals
J6527406 |
5 g |
Each for $867.64
|
|
|||||
Description
Inhibits bacterial cell wall synthesis
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
27164-46-1 | |
479.504 | |
MTIAAUXSENDLGW-SLNAEPSVSA-N | |
131673922 | |
[HH].CC1=NN=C(S1)SCC2=C(N3C(C(C3=O)NC(=O)CN4C=NN=N4)SC2)C(=O)O.[Na] |
C14H16N8NaO4S3 | |
MFCD00056883 | |
Cefamedin | |
(6R,7R)-3-[(5-methyl-1,3,4-thiadiazol-2-yl)sulfanylmethyl]-8-oxo-7-[[2-(tetrazol-1-yl)acetyl]amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;molecular hydrogen;sodium |
Specifications
Cefazolin Sodium Salt | |
27164-46-1 | |
C14H16N8NaO4S3 | |
3585038 | |
Cefamedin | |
[HH].CC1=NN=C(S1)SCC2=C(N3C(C(C3=O)NC(=O)CN4C=NN=N4)SC2)C(=O)O.[Na] | |
479.504 | |
476.48 | |
1 g | |
Cefazolin sodium salt |
Powder | |
170°C to 172°C | |
MFCD00056883 | |
14,1917 | |
MTIAAUXSENDLGW-SLNAEPSVSA-N | |
(6R,7R)-3-[(5-methyl-1,3,4-thiadiazol-2-yl)sulfanylmethyl]-8-oxo-7-[[2-(tetrazol-1-yl)acetyl]amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;molecular hydrogen;sodium | |
131673922 | |
White | |
Light sensitive |
Safety and Handling
GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c
H317-H334
EINECSNumber : 248-278-4
RTECSNumber : XI0390000
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only