missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ceftezole 98.0+%, TCI America™

Supplier:  TCI America C277025G

 View more versions of this product

Catalog No. C277025G

Explore available promotions

Description

Description

Specifications

Specifications

Ceftezole
163°C
C13H12N8O4S3
25 g
C1C(=C(N2C(S1)C(C2=O)NC(=O)CN3C=NN=N3)C(=O)O)CSC4=NN=CS4
440.471
440.47
Crystalline Powder
26973-24-0
Yellow
MFCD00274176
DZMVCVMFETWNIU-LDYMZIIASA-N
(6R,7R)-8-oxo-7-[[2-(tetrazol-1-yl)acetyl]amino]-3-(1,3,4-thiadiazol-2-ylsulfanylmethyl)-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
65755
≥98.0% (HPLC,T)

Chemical Identifiers

26973-24-0
440.471
DZMVCVMFETWNIU-LDYMZIIASA-N
(6R,7R)-8-oxo-7-[[2-(tetrazol-1-yl)acetyl]amino]-3-(1,3,4-thiadiazol-2-ylsulfanylmethyl)-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
C13H12N8O4S3
MFCD00274176
65755
C1C(=C(N2C(S1)C(C2=O)NC(=O)CN3C=NN=N3)C(=O)O)CSC4=NN=CS4
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions