missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Cel-Gro Tissue Culture Rotator Drums

Find the right tissue culture rotator drum to hold test tubes of various sizes with these rotator accessory options.

Supplier:  Thermo Scientific™ 1651Q

 View more versions of this product

Catalog No. 14253100Q


Explore available promotions

Description

Description

Gain versatility to carry out a variety of rotating and mixing operations with Thermo Scientific™ Cel-Gro Tissue Culture Rotator Drums, which are accessories for rotators. Achieve optimum culture results for tissue and virus growth, hormone production, sample testing to determine the presence of viruses and cytotoxicity assays, and other applications with the right rotator accessories.

  • The variable-speed feature permits a range of motion from a slow, gentle rotation, 12 rpm, to a progression of more vigorous rotating speeds up to 70 rpm.
  • Drums must be ordered separately. The drums consist of 3 circular plates (2 are perforated) that are carefully aligned and securely formed into a rigid assembly of heavy-gauge, anodized aluminum.
  • Can be operated in standard incubators for cultivation under controlled temperature conditions.
  • A pilot light indicates when units are operational.
  • Every unit is finished in a powder coated stainless steel for easy clean up and maintenance.

Warranty: 90 days

Specifications

Specifications

30 mm
Drum: 14 in.
48.26 x 48.26 x 20.32 cm
60
Rotator Drums
Drum: 36 cm
Rotator
Videos
SDS
Documents

Documents

Promotions

Promotions

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.