Learn More
Celestine Blue, pure
CAS: 1562-90-9 | C17H18ClN3O4 | 363.798 g/mol
$242.35 - $811.08
Chemical Identifiers
| CAS | 1562-90-9 |
|---|---|
| Molecular Formula | C17H18ClN3O4 |
| Molecular Weight (g/mol) | 363.798 |
| MDL Number | MFCD00011927 |
| InChI Key | OJAMERVLRXYMPD-UHFFFAOYSA-N |
| PubChem CID | 54684697 |
| IUPAC Name | 7-(diethylamino)-4-hydroxy-3-oxophenoxazin-10-ium-1-carboxamide;chloride |
| SMILES | CCN(CC)C1=CC2=C(C=C1)[NH+]=C3C(=CC(=O)C(=C3O2)O)C(=O)N.[Cl-] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC195560100
|
Thermo Scientific Chemicals
195560100 |
10 g |
Each for $242.35
|
|
|||||
|
AC195560500
|
Thermo Scientific Chemicals
195560500 |
50 g |
Each for $811.08
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1562-90-9 | |
| 363.798 | |
| OJAMERVLRXYMPD-UHFFFAOYSA-N | |
| 7-(diethylamino)-4-hydroxy-3-oxophenoxazin-10-ium-1-carboxamide;chloride |
| C17H18ClN3O4 | |
| MFCD00011927 | |
| 54684697 | |
| CCN(CC)C1=CC2=C(C=C1)[NH+]=C3C(=CC(=O)C(=C3O2)O)C(=O)N.[Cl-] |
Specifications
| 1562-90-9 | |
| MFCD00011927 | |
| OJAMERVLRXYMPD-UHFFFAOYSA-N | |
| CCN(CC)C1=CC2=C(C=C1)[NH+]=C3C(=CC(=O)C(=C3O2)O)C(=O)N.[Cl-] | |
| 363.798 | |
| 638 to 648nm | |
| Extra Pure | |
| 700 min. (in water) | |
| Black to Brown | |
| Powder |
| C17H18ClN3O4 | |
| 15, 1961 | |
| Authentic | |
| 7-(diethylamino)-4-hydroxy-3-oxophenoxazin-10-ium-1-carboxamide;chloride | |
| 54684697 | |
| 363.79 | |
| Glass bottle | |
| Complies | |
| 10 g | |
| Celestine blue |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause respiratory irritation.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
RUO – Research Use Only