missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Celiprolol hydrochloride, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals J65943MA
Description
A cardioselective 1-adrenergic blocker
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Celiprolol hydrochloride | |
White | |
MFCD00941500 | |
14,1960 | |
Soluble in ethanol, methanol and water | |
[H+].[Cl-].CCN(CC)C(=O)NC1=CC=C(OCC(O)CNC(C)(C)C)C(=C1)C(C)=O | |
415.96 | |
415.95 |
57470-78-7 | |
C20H34ClN3O4 | |
10 mg | |
celiprolol hydrochloride, celiprolol hcl, selectrol, selecor, selectol tn, celectol, ccris 1095, celiprolol hydrochloride usan, celiprolol clorhidrato spanish, celiprolol hydrochlorid german | |
VKJHTUVLJYWAEY-UHFFFAOYNA-N | |
3-[3-acetyl-4-[3-(tert-butylamino)-2-hydroxypropoxy]phenyl]-1,1-diethylurea;hydrochloride | |
42373 | |
Crystalline Solid |
Chemical Identifiers
57470-78-7 | |
415.96 | |
VKJHTUVLJYWAEY-UHFFFAOYNA-N | |
42373 | |
[H+].[Cl-].CCN(CC)C(=O)NC1=CC=C(OCC(O)CNC(C)(C)C)C(=C1)C(C)=O |
C20H34ClN3O4 | |
MFCD00941500 | |
celiprolol hydrochloride, celiprolol hcl, selectrol, selecor, selectol tn, celectol, ccris 1095, celiprolol hydrochloride usan, celiprolol clorhidrato spanish, celiprolol hydrochlorid german | |
3-[3-acetyl-4-[3-(tert-butylamino)-2-hydroxypropoxy]phenyl]-1,1-diethylurea;hydrochloride |
Safety and Handling
EINECSNumber : 260-752-2
RTECSNumber : YR6560000
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only