Cell Culture Grade Water Tested to USP/EP Sterile Water, Corning™
High quality water for use in cell culture applications. Corning™ Cell Culture Grade Water Tested to USP/EP Sterile Water are used in preparation of purification and chromatography buffers.
$594.24 - $2554.48
Specifications
Quantity | 20 L |
---|
Alert:
Product should not be used as an injectable or in the preparation of injectables.
Description
Cell culture grade water is high-quality water suitable for use as a solvent in the preparation of cell culture media and other reagents for sensitive applications. Our bioprocess container selections are suitable for large-scale applications such as cleaning and upstream and downstream reagent preparation. Cell culture grade water goes through a wide array of testing which includes testing to the chemical and physical properties found in the USP and EP monographs for sterile Water for Injection (WFI).
Certifications
Manufactured under cGMP (21 CFR 820) in ISO 13485 certified facility
Specifications
20 L |