Greiner Bio-One CELLdisc Advanced Multilayer Cell Culture Surface
CELLdisc multilayer device for simplifying scale up and mass cell culture. Covers a range of cell culture surfaces from 250 cm2 up to 10,000 cm2
Supplier:  Greiner Bio-One 678916
 View more versions of this product
Catalog No. 07-001-062
Explore available promotions
Description
Description
  • New Advanced TC treated multilayer device covering a range of cell culture surfaces from 1000 cm2 to 10,000 cm2
  • Minimum space required for handling of device 40% higher surface/volume ratio than conventional multilayer systems
  • Advanced TC Surface treatment for optimal cell attachment
  • Optimal ventilation through central gas support channel
  • Gas inlet and outlet equipped with filters
  • Easy accessibility due to wide opening port
  • Predictable scale up within one format (1000 cm2 to 10,000 cm2)
  • Suitable for basic research as well as industrial applications
Specifications
Specifications
Blue
214.8 mm
4° to 37°C
4000 cm²
1
Polystyrene
Cell Culture Flask
Screw Cap
16