Thermo Scientific™ CellInsight™ CX7 LZR Pro High Content Screening Platform DFS Item
The Thermo Scientific CellInsight CX7 LZR Pro High Content Screening (HCS) Platform is a fast, laser light engine-based, automated cellular imaging and analysis platform designed for life scientists who require on-the-fly quantitative analysis.
Supplier:  Thermo Scientific™ HCSDCX7LZRPRO
Catalog No. 50-272-3662
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
  • A fast, laser light engine-based, automated cellular imaging and analysis platform designed for life scientists who require on-the-fly quantitative analysis
  • The CellInsight CX7 LZR Pro Platform offers a more powerful and integrated way to develop and automate high content assays using a combination of 5-color bright-field and 7-color fluorescence imaging in confocal or wide-field modes
  • With laser illumination, the CellInsight LZR Pro Platform enables researchers to image faster in both wide-field and confocal mode
Features of the CellInsight CX7 LZR Pro Platform:
  • Back-illuminated sCMOS camera offers a 95% peak quantum efficiency and -20°C cooling
  • New validated applications including Cell Painting and FirePlex-HT immunoassay
  • Olympus X-line objective imaging to improve imaging quality and assay performance
  • EurekaScan Finder double-pass functionality for on-the-fly seek and find of spheroids, tissue, and rare cellular events
  • HCS Studio 5.0 with over 40 life science bio-applications and real-time analysis for fastest time-to-data possible.
  • 7-channel fluorescent imaging with laser illumination
  • 5-channel bright-field imaging
  • 7-channel confocal imaging with laser illumination
  • Software and laser-based autofocus for consistent scan times
  • Configured to accept optional modules for fully automated plate handling and live cell on-board environmental control
Capabilities
  • Use integrated HCS Studio software for instrument control and data analysis
  • Achieve the fastest time to results by using intelligent acQuisition (iQ) real-time analysis feedback
  • Addresses hundreds of assay types with flexible software and hardware tools
  • Scan and analyze microscope slides or most SBS-standard format microplates, including 6-well through 1536-well formats
  • Compatible with Molecular Probes fluorescent detection reagents
Hardware and software
  • High-output laser excitation solid state light
  • Broad reagent options with 7-color excitation from violet to near IR for assay multiplexing
  • Dual mode focusing using Software and Laser Autofocus methods to address both labelled and unlabeled sample types
  • Multi-mode imaging allows for wide-field, bright-field, and confocal imaging in one run
  • HCS Studio 5.0 Cell Analysis Software is an integrated instrument control, visualization, and analysis software package for novice to advanced users
  • Scalable with Store Image and Database Software and optional Orbitor RS Microplate handler (not included)
Specifications
Specifications
CellInsight
Highly Sensitive Monochrome CMOS
0 to 3000 VDC, 0 to 250 VAC
Linear, Orbital, Double Orbital
1 each
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only.