missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S'il vous plaît vous connecter à votre compte en ligne pour afficher votre prix réduits
Thermo Scientific™ CellInsight™ CX7 LED Pro High Content Screening Platform
The Thermo Scientific CellInsight CX7 Pro High Content Screening (HCS) Platform is a fast, LED engine-based, automated cellular imaging and analysis platform designed for life scientists whom require on-the-fly quantitative analysis.
Supplier: Thermo Scientific™ HCSDCX7LEDPRO
Description
- A fast, LED engine-based, automated cellular imaging and analysis platform designed for life scientists whom require on-the-fly quantitative analysis.
- The CellInsight CX7 Pro HCS Platform offers an integrated way to develop and automate high content assays using a combination of 5-color brightfield and 7-color fluorescence imaging in confocal or wide-field modes
Features of the CellInsight CX7 Pro Platform
- Back-illuminated sCMOS camera offer a 95% peak quantum efficiency and -20°C cooling
- New validated applications including Cell Painting and FirePlex-HT immunoassay
- Olympus X-line objective imaging to improve imaging quality and assay performance
- EurekaScan Finder double-pass functionality for on-the-fly seek and find of spheroids, tissue, and rare cellular events
- HCS Studio 5.0 with over 40 life science bio-applications and real-time analysis for fastest time-to-data possible
- 7-channel fluorescent imaging
- 5-channel bright-field imaging
- 7-channel confocal imaging
- Software and laser-based autofocus for consistent scan times
- Configured for fully automated plate handling and scanning
Capabilities
- Use HCS Studio software for integrated instrument control and data analysis
- Achieve the fastest time to results by using intelligent acQuisition (iQ) real-time analysis feedback
- Addresses hundreds of assay types with flexible software and hardware tools
- Scan and analyze microscope slides or most SBS-standard format microplates
- Compatible with Molecular Probes fluorescent detection reagents
Hardware and software
- Stable and reliable light source enables reduced variability, increased data quality, and decreased operating costs
- Broad reagent options with 7-color excitation from violet to near IR for assay multiplexing
- Dual mode focusing using Software and Laser Autofocus methods to address both labelled and unlabeled sample types
- Multi-mode imaging allows for wide-field, bright-field, and confocal imaging in one run
- HCS Studio 5.0 Cell Analysis Software is an integrated instrument control, visualization, and analysis software package for novice to advanced users
- Scalable with Store Image and Database Software and optional Orbitor RS Microplate handler (not included)
Spécifications
CellInsight | |
Highly Sensitive Monochrome CMOS | |
0 to 3000 VDC, 0 to 250 VAC |
Linear, Orbital, Double Orbital | |
1 each |
For Research Use Only.