Molecular Probes™ CellROX™ Orange Flow Cytometry Assay Kit DFS Item
CellROX™ Orange Flow Cytometry Assay Kit
Supplier:  Molecular Probes™ C10493
Catalog No. 501121510
Explore available promotions
Description
Description
The CellROX™ Orange Flow Cytometry Assay Kit enables the flow cytometric detection of reactive oxygen species (ROS) in live cells. The kit includes the novel, fluorogenic CellROX™ Orange Reagent, as well as SYTOX™ Red Dead Cell Stain, N-acetyl cysteine (an antioxidant, for negative control), and tert-butyl hydroperoxide solution (TBHP, an inducer of ROS).
View a selection guide for all CellROX™ Reagents and Kits.
The CellROX™ Orange Flow Cytometry Assay Kit features:
  • Fluorogenic probe formulated for flow cytometry that is oxidized in the presence of ROS
  • Multicolor compatibility—minimal overlap with fluorophores excited by other laser lines, allowing easy multiplexing with other reagents
  • Simple protocol—cells can be stained in complete media or other appropriate buffer, no need for serum free media
    The CellROX™ Orange Detection Reagent is cell-permeable and non-fluorescent or very weakly fluorescent while in the reduced state. Upon oxidation, the reagent exhibits a strong fluorogenic signal that has absorption/emission maxima of 545/565 nm and remains localized in the cytoplasm. When used together with the included SYTOX™ Red Dead Cell Stain, oxidatively stressed and nonstressed cells are reliably distinguished from dead cells by flow cytometry.
  • Specifications
    Specifications
    Tube
    100 assays
    CellROX orange: 545/565, SYTOX red: 640/658
    Videos
    SDS
    Documents
    Documents
    Promotions
    Promotions
    For Research Use Only. Not for use in diagnostic procedures.