missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Greiner Bio-One CELLSTAR 96-well, Cell Culture-Treated, Flat-Bottom, Half-Area Microplate

Color:Black; Sterility:Sterile; Lid:With Lid

Supplier:  Greiner Bio-One 675086

Catalog No. 07000095


Explore available promotions

Description

Description

Alphanumeric coding
Specifications

Specifications

Half-Area
Fluorescence
Not Autoclavable
Polystyrene
15 to 175 μL
Cell Culture
Round, Flat Bottom (F-well)
4800 x g (swinging bucket rotor)
8
Yes
Chimney
With Lid
8 x 12
Black
96
Sterile
199 μL
Yes
32
32/Cs.
Black
127.76 x 85.48 x 14.40 mm
SDS
Documents

Documents

Promotions

Promotions