Learn More
Greiner Bio-One CELLSTAR Standard TC Treated Cell Culture Flask with Measuring Grid
Improved cell adhesion through physical surface treatment. Greiner Bio-One CELLSTAR Standard TC Treated Cell Culture Flask with Measuring Grid is free of detectable DNase, RNase and human DNA.
Supplier:  Greiner Bio-One 690170
Catalog No. 07000230
Only null left
Explore available promotions
Description
Description
  • With measuring grid
  • Allows efficient access to the cell lawn with cell scrapers and optimal access with pipettes
  • Graduation on the side
  • Canted neck style
  • Standard plug cap
Specifications
Specifications
Red
50 mL
Canted
10
Yes
Standard screw cap
HDPE
Polystyrene
200
Physical Surface Treatment
Sterile
Product Suggestions
Product Suggestions