Promega CellTiter-Glo™ 2.0 Assay DFS Item
The CellTiter-Glo™ 2.0 Assay provides a homogeneous method of determining the number of viable cells in culture by quantitating the amount of ATP present, which indicates the presence of metabolically active cells.
Supplier:  Promega G9242
 View more versions of this product
Catalog No. PRG9242
Explore available promotions
Description
Description
  • A Luminescent Cell Viability Assay for Fast, Easy, Everyday Use
  • Reduced preparation time
  • Convenient repeat use over multiple days
  • Same great performance as classic CellTiter-Glo(R) Assay
Specifications
Specifications
100 mL