missing translation for 'onlineSavingsMsg'
Learn More

Cellulose Acetate Butyrate, Butyryl Content 50-54%, Thermo Scientific™

Chemical Identifiers

CAS 9004-36-8
Molecular Formula (C12H14O10)n
MDL Number MFCD00081513
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC404225000
SDS View Documents
Thermo Scientific Chemicals
404225000
500g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC404220025
SDS View Documents
Thermo Scientific Chemicals
404220025
2.5kg Plastic drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

9004-36-8
(C12H14O10)n
MFCD00081513

Specifications

9004-36-8
(C12H14O10)n
500g
Glass bottle
0.01 to 0.03 (sec.)
16000 (Mn)
MFCD00081513
Authentic
1.475
Cellulose acetate butyrate, Butyryl Content 50-54%
Videos
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only