missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals™ CellVation™ Cryopreservation Medium

Recommended for the preservation of stem cells, hepatocytes, tissue samples, and a wide variety of cell types and established cell lines

Supplier:  MP Biomedicals™ 0920300M2

 View more versions of this product

Catalog No. MP920300M2


Explore available promotions

Description

Description

CellVation is a cryopreservation medium designed for optimal cell recovery and convenience. It does not contain DMSO or serum, and is scientifically engineered for serum-free cultures, as well as, ideally suited for all types of serum. It has also been shown to preserve the potential of stem cells for differentiation.

Specifications

Specifications

Mycoplasma, Bacteria, Fungi, Yeast and Mold free. Certified U.S. origin.
CellVation Cryopreservation Media
Cryopreservation Media
Cell counting, viability and cryopreservation
60 mL
Store at 4°C
Solution
<0.5 EU/mL
6.5 to 7.5
Videos
SDS
Documents

Documents

Promotions

Promotions

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.