missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Eppendorf™ CellXpert™ CS220 - Large Capacity CO2 Incubator Shaker

Accelerate your research with the Eppendorf™ CellXpert™ CS220 CO2 Incubator Shaker - the only CO2 incubator shaker with integrated 180°C sterilization. Achieve high flask capacity and reliable results with optimized lab space efficiency and low operating costs.

Supplier:  Eppendorf™ 6740112030

 View more versions of this product

Catalog No. 01-670-843

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Accelerate your growth - CellXpert™ CS220 - CO2 Incubator Shaker

The CellXpert™ CS220 is a large capacity, stackable CO2 Incubator Shaker that was designed to accelerate mammalian suspension culture research projects.

Please order separately: platform, clamps, sticky pads, storage base or feet, water supply package. Optional: 1-2 stacking kits and water tank holder

  • High flask capacity
  • 180°C sterilization routine integrated: including wizard and downloadable performance protocol, no locally restricted UV-airflow/surface-disinfection
  • Stainless steel chamber: Hygienic design according to EN ISO 14159:2008 standard plus only few internal parts and no drive wheel, crevices, cables or heating fins inside
  • Optimized throughput per lab space: Highest platform size vs. footprint ratio in the market, stackable up to 3-fold
  • Saves costs: Always included: slide-out platform, rH-control, touch interface, access port, 180°C sterilization routine, no UV-lamps that require frequent exchange, low gas and energy consumption
  • Eppendorf X-drive: Proven and tested shaker drive with four weight-supporting shafts for 24/7 vibration-free shaking
  • Glide-up door and slide-out platform: Saves space and ensures fast and easy experiment setup, cleaning and service – even at the back of the chamber
  • Full control and fast root-cause analysis: 7 in. touchscreen shows all setpoints and current parameters at a glance, direct visual overview of logged data/events including door openings and alarms, user management

Applications:

  • Protein Expression
  • Screening
  • Cell line development
  • Early process development
  • Viral vector production in mammalian cells e.g., Ad, AAV, LV
  • Bioreactor inoculum preparation
Specifications

Specifications

51 mm
20 to 250 rpm
8°C above ambient to 60°C
Incubator Shaker
0.1°C increments
50 to 60 Hz
22.8 in.
20.2 x 36.6 in.
237 kg
CO2 Range: 0.1% to 20% CO2, CO2 Sensor type: NDIR
Color Touch Screen
Eppendorf X-drive
No
18 kg
100/130 V
8 x 5 L Corning™ and Thomson Optimum Growth™ (max. flask size), 25 x 1 L Erlenmeyer flasks, 42 x 500 mL Erlenmeyers flasks, 102 x mL Erlenmeyer flasks
± 0.4°C at 37°C
± 0.3°C at 37°C
31.7 in.
22.8 x 40.4 x 15.2 in.
522.5 lb.
CO2 Accuracy: ± 0.3% at 5% CO2
CellXpert CS220
Active bidirection humidity control up to 85% relative humidity, non-condensing
224 L
Videos
SDS
Documents

Documents

Promotions

Promotions