missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
CEP Cylinder Dolly HD
Transport cylinders in industrial or outdoor situations
Supplier: CEP 7301CG
Description
- Accommodates Class 300 SCF industrial gas cylinders and No. 5 acetylene cylinders
- Constructed from durable polyethylene
- Single cylinder
- Load capacity: 113.4kg (250 lb.)
- Size: 57 × 65 × 117cm (22.5 × 24.75 × 46 in.)
For demanding industrial facilities or tough outdoor applications in which transport of cylinders is required.
Specifications
Cylinder Dolly HD | |
113.4 kg | |
Industrial facilities | |
117 cm | |
57 cm | |
65 cm |
250 lb. | |
22.5 x 24.75 x 46 in. (57 x 65 x 117 cm) | |
46 in. | |
22.5 in. | |
24.75 in. |