CEP Universal Aggressive Sorbent Pads and Rolls
Absolute necessity for any Haz Mat spill or spill contingency planning
Supplier:  CEP UP50
 View more versions of this product
Catalog No. 19-056-212
Explore available promotions
Description
Description
  • Absorb all liquids from water-based fluids to aggressive acids
  • Bonded for tensile strength
  • Absorb up to 25 times their weight in all liquids
Specifications
Specifications
Gray
166.5 L
Any hazmat spill or spill contingency planning
30 x 30 in. (76.2 x 76.2 cm)
76.2 cm
76.2 cm
44 gal.
Universal Aggressive Sorbent Pad
Heavyweight
30 in.
30 in.