CEP Universal Aggressive Sorbent Pads and Rolls
Absolute necessity for any Haz Mat spill or spill contingency planning
Supplier:  CEP USR150
 View more versions of this product
Catalog No. 19-056-214
Explore available promotions
Description
Description
  • Absorb all liquids from water-based fluids to aggressive acids
  • Bonded for tensile strength
  • Absorb up to 25 times their weight in all liquids
Specifications
Specifications
Gray
189 L
Any hazmat spill or spill contingency planning
150 ft. x 14 in. (45.7 m x 35.6 cm)
45.7 m
35.6 cm
50 gal.
Universal Aggressive Sorbent Roll
Heavyweight
150 ft.
14 in.