missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cephalexin Monohydrate USP MP Biomedicals
Supplier: MP Biomedicals Inc 0215058525
Specifications
Cephalexin Monohydrate | |
Crystalline Powder | |
Beige to White | |
≥95% | |
MFCD00167148,MFCD00056877 | |
AVGYWQBCYZHHPN-CYJZLJNKSA-N | |
(6R,7R)-7-[(2R)-2-amino-2-phenylacetamido]-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid hydrate | |
62921 | |
365.4 |
Cephalexin Monohydrate | |
23325-78-2 | |
25 g | |
C16H19N3O5S | |
7-(d-a-Aminophenylacetamido)desacetoxycephalosporanic acid, Cefalexin monohydrate | |
O.CC1=C(N2[C@H](SC1)[C@H](NC(=O)[C@H](N)C1=CC=CC=C1)C2=O)C(O)=O | |
365.40 | |
CHEBI:3535 | |
BP/USP |
Chemical Identifiers
23325-78-2 | |
365.40 | |
AVGYWQBCYZHHPN-CYJZLJNKSA-N | |
62921 | |
(6R,7R)-7-[(2R)-2-amino-2-phenylacetamido]-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid hydrate |
C16H19N3O5S | |
MFCD00167148,MFCD00056877 | |
7-(d-a-Aminophenylacetamido)desacetoxycephalosporanic acid, Cefalexin monohydrate | |
CHEBI:3535 | |
O.CC1=C(N2[C@H](SC1)[C@H](NC(=O)[C@H](N)C1=CC=CC=C1)C2=O)C(O)=O |
Safety and Handling
EINECSNumber : 239-773-6
Recommended Storage : Store at 4°C, desiccate.