missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cephalothin 98.0+%, TCI America™

Supplier:  TCI America C27695G

 View more versions of this product

Catalog No. C27695G


Explore available promotions

Description

Description

Specifications

Specifications

Cephalothin
154°C
C16H15N2NaO6S2
5 g
VUFGUVLLDPOSBC-XRZFDKQNSA-M
sodium (6R,7R)-3-[(acetyloxy)methyl]-8-oxo-7-[2-(thiophen-2-yl)acetamido]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
6024
396.43
Crystalline Powder
153-61-7
Yellow
MFCD00242614
Cephalotin, 7-(2-Thienylacetamido)cephalosporanic Acid
[Na+].CC(=O)OCC1=C(N2[C@H](SC1)[C@H](NC(=O)CC1=CC=CS1)C2=O)C([O-])=O
418.41
CHEBI:124991
≥98.0% (HPLC,T)

Chemical Identifiers

153-61-7
418.41
VUFGUVLLDPOSBC-XRZFDKQNSA-M
6024
sodium (6R,7R)-3-[(acetyloxy)methyl]-8-oxo-7-[2-(thiophen-2-yl)acetamido]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
C16H15N2NaO6S2
MFCD00242614
Cephalotin, 7-(2-Thienylacetamido)cephalosporanic Acid
CHEBI:124991
[Na+].CC(=O)OCC1=C(N2[C@H](SC1)[C@H](NC(=O)CC1=CC=CS1)C2=O)C([O-])=O
Videos
Safety and Handling

Safety and Handling

RTECSNumber : XI0388000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions